1,8-Naphthalenedicarboxylic anhydride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

N0022-25G N0022-250G
TCN0022-025GEA 48.62 CAD
TCN0022-025G TCN0022-250G
1,8-Naphthalenedicarboxylic anhydride ≥98.0% (by HPLC, titration analysis)
1,8-Naphthalenedicarboxylic anhydride

CAS Number: 81-84-5
MDL Number: MFCD00006925
Molecular Formula: C12H6O3
Molecular Weight: 198.18
Purity/Analysis Method: >98.0% (T,HPLC)
Form: Crystal
Melting point (°C): 274


Other Product Names / Synonyms: 1,8-Naphthalenedicarboxylic Anhydride

Formula: C₁₂H₆O₃
Boiling Pt: 425 °C (1013 hPa)
Melting Pt: 274 °C
MDL Number: MFCD00006925
CAS Number: 81-84-5
EINECS: 201-380-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae47420e3b064fcb7f82582bf79aa086503df716&vid=ae47420e3b064fcb7f82582bf79aa086503df716&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR