2-Naphthaldehyde ≥98.0%

Supplier: TCI America

N0003-5G N0003-25G
TCN0003-005GEA 70.44 CAD
TCN0003-005G TCN0003-025G
2-Naphthaldehyde ≥98.0%
2-Naphthaldehyde

CAS Number: 66-99-9
MDL Number: MFCD00004094
Molecular Formula: C11H8O
Molecular Weight: 156.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 147
Melting point (°C): 60

Formula: C₁₁H₈O
Boiling Pt: 147 °C
Melting Pt: 60 °C
Density: 1.0775 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004094
CAS Number: 66-99-9
EINECS: 200-640-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a90f3b734d3dfcdef45ee4f1822e8b5b58b59990&vid=a90f3b734d3dfcdef45ee4f1822e8b5b58b59990&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR