2'-Methoxyacetophenone ≥97.0%

Supplier: TCI America

M1445-25G M1445-250G
TCM1445-25GEA 143.78 CAD
TCM1445-25G TCM1445-250G
2'-Methoxyacetophenone ≥97.0%
2'-Methoxyacetophenone

CAS Number: 579-74-8
MDL Number: MFCD00008725
Molecular Formula: C9H10O2
Molecular Weight: 150.18
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 108
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: 2-Acetoanisole, 2-Acetylanisole

Formula: C₉H₁₀O₂
Boiling Pt: 131 °C (1013 hPa)
Density: 1.09 (20 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00008725
CAS Number: 579-74-8
EINECS: 209-446-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab78d9d1724f31d809047731b64bab1745d0652d&vid=ab78d9d1724f31d809047731b64bab1745d0652d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR