5-Methoxyisatin ≥95.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

M1362-25G M1362-5G
TCM1362-25GEA 886.38 CAD
TCM1362-25G TCM1362-005G
5-Methoxyisatin ≥95.0% (by HPLC, titration analysis)
5-Methoxyisatin

CAS Number: 39755-95-8
MDL Number: MFCD00169023
Molecular Formula: C9H7NO3
Molecular Weight: 177.16
Purity/Analysis Method: >95.0% (HPLC,T)
Form: Crystal
Melting point (°C): 203


Product Categories: Synthetic Organic Chemistry, Isatins (Multi-component Reaction)
Other Product Names / Synonyms: 5-Methoxyindole-2,3-dione, 5-Methoxy-2,3-indolinedione

Formula: C₉H₇NO₃
Melting Pt: 203 °C
MDL Number: MFCD00169023
CAS Number: 39755-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a116ffbd04bc1bc453cc9a1fbf1449574a40bde9&vid=a116ffbd04bc1bc453cc9a1fbf1449574a40bde9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR