2-Mercapto-5-methylbenzimidazole

Supplier: TCI America

M1357-100G M1357-25G
TCM1357-100GEA 200.66 CAD
TCM1357-100G TCM1357-25G
2-Mercapto-5-methylbenzimidazole
2-Mercapto-5-methylbenzimidazole

CAS Number: 27231-36-3
MDL Number: MFCD00010617
Molecular Formula: C8H8N2S
Molecular Weight: 164.23
Form: Crystal
Color: White


Other Product Names / Synonyms: 5-Methyl-2-benzimidazolethiol

Formula: C₈H₈N₂S
Boiling Pt: 349 °C (1013 hPa)
Melting Pt: 220 °C
MDL Number: MFCD00010617
CAS Number: 27231-36-3
EINECS: 248-350-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=de25843ddac1fde76daada5c2eb605838522266b&vid=de25843ddac1fde76daada5c2eb605838522266b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR