(S)-(+)-Methyl mandelate ≥98.0%

Supplier: TCI America

Synonyms: L(+)-Mandelic acid methyl ester, L(+)-2-Hydroxy-2-phenylacetic acid methyl ester, Methyl (S)-(+)-mandelate, L(+)-Methyl mandelate, (S)-(+)-Mandelic acid methyl ester, (S)-(+)-2-Hydroxy-2-phenylacetic acid methyl ester, L(+)-Methyl glycolate, (S)-(+)-Methyl glycolate

M1350-5G M1350-1G
TCM1350-005GEA 37.92 CAD
TCM1350-005G TCM1350-001G
(S)-(+)-Methyl mandelate ≥98.0%
(S)-(+)-Methyl mandelate

CAS Number: 21210-43-5
MDL Number: MFCD00064246
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 138
Melting point (°C): 55
Flash Point (°C): 110
Specific rotation [a]20/D: 140 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Mandelic Acid Methyl Ester

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 138 °C
Melting Pt: 58
Flash Pt: 110 °C
MDL Number: MFCD00064246
CAS Number: 21210-43-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cd2c0ffe0370228598fe77879e4e1962dfd8b87&vid=7cd2c0ffe0370228598fe77879e4e1962dfd8b87&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR