(R)-(-)-Methyl mandelate ≥98.0%

Supplier: TCI America

Synonyms: D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester

M1349-5G M1349-1G
TCM1349-005GEA 47.4 CAD
TCM1349-005G TCM1349-001G
(R)-(-)-Methyl mandelate ≥98.0%
(R)-(-)-Methyl mandelate

CAS Number: 20698-91-3
MDL Number: MFCD00064247
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Specific rotation [a]20/D: -145 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: D-(-)-Mandelic Acid Methyl Ester

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 51…53 °C
Flash Pt: 110 °C
MDL Number: MFCD00064247
CAS Number: 20698-91-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=568f5741483ae65461ade5753835d7e0cf09def5&vid=568f5741483ae65461ade5753835d7e0cf09def5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR