6-Methoxyflavone ≥99.0%

Supplier: TCI America

M1346-1G M1346-5G
TCM1346-001GEA 93.22 CAD
TCM1346-001G TCM1346-005G
6-Methoxyflavone ≥99.0%
6-Methoxyflavone

CAS Number: 26964-24-9
MDL Number: MFCD00017322
Molecular Formula: C16H12O3
Molecular Weight: 252.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 165

Formula: C₁₆H₁₂O₃
Melting Pt: 165 °C
MDL Number: MFCD00017322
CAS Number: 26964-24-9
EINECS: 248-144-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=093c865b7324d4d5e4fedac2194d071d38b92cb8&vid=093c865b7324d4d5e4fedac2194d071d38b92cb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR