3-Methoxyphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
M1322-5G
M1322-1G
M1322-25G
TCM1322-5GEA
71.1
CAD
TCM1322-5G
TCM1322-001G
TCM1322-25G
3-Methoxyphenylboronic acid (contains varying amounts of Anhydride)
3-Methoxyphenylboronic acid
CAS Number: 10365-98-7
MDL Number: MFCD00161359
Molecular Formula: C7H9BO3
Molecular Weight: 151.96
Form: Crystal
Color: White
Melting point (°C): 160
Other Product Names / Synonyms: 3-Methoxybenzeneboronic Acid
Formula:
C₇H₉BO₃ Melting Pt: 160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00161359 CAS Number: 10365-98-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...