3-Tolylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

M1314-25G M1314-1G M1314-5G
TCM1314-25GEA 366.56 CAD
TCM1314-25G TCM1314-001G TCM1314-005G
3-Tolylboronic acid (contains varying amounts of Anhydride)
3-Tolylboronic acid

CAS Number: 17933-03-8
MDL Number: MFCD00040198
Molecular Formula: C7H9BO2
Molecular Weight: 135.96
Form: Crystal
Color: White


Other Product Names / Synonyms: 3-Methylbenzeneboronic Acid, m-Tolylboronic Acid

Formula: C₇H₉BO₂
Melting Pt: 165 °C
MDL Number: MFCD00040198
CAS Number: 17933-03-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12296523c6c19989b00dd1755e564c62154b0df6&vid=12296523c6c19989b00dd1755e564c62154b0df6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR