5-Methoxyindol-3-ylacetic acid ≥98.0%

Supplier: TCI America

Synonyms: 5-Methoxy-3-indoleacetic acid, 2-(5-Methoxy-1H-indol-3-yl)acetic acid, 5-Methoxyindole-3-acetic acid

M1303-5G M1303-1G
TCM1303-005GEA 1431.48 CAD
TCM1303-005G TCM1303-001G
5-Methoxyindol-3-ylacetic acid ≥98.0%
5-Methoxyindol-3-ylacetic acid

CAS Number: 3471-31-6
MDL Number: MFCD00005638
Molecular Formula: C11H11NO3
Molecular Weight: 205.21
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 148

Formula: C₁₁H₁₁NO₃
MW: 205.21 g/mol
Melting Pt: 148 °C
MDL Number: MFCD00005638
CAS Number: 3471-31-6
EINECS: 222-438-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f7a73e92524fbcdcc89709511074da297e82478&vid=3f7a73e92524fbcdcc89709511074da297e82478&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR