3-Methylcinnamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M1298-25G M1298-5G
TCM1298-025GEA 562.48 CAD
TCM1298-025G TCM1298-005G
3-Methylcinnamic acid ≥98.0% (by titrimetric analysis)
3-Methylcinnamic acid

CAS Number: 3029-79-6
MDL Number: MFCD00016844
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 116
Flash Point (°C): 110

Formula: C₁₀H₁₀O₂
Boiling Pt: 303 °C (1013 hPa)
Melting Pt: 116 °C
Flash Pt: 110 °C
MDL Number: MFCD00016844
CAS Number: 3029-79-6
EINECS: 221-199-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56ff02c19ce1371a2a49ed5e7fc9af49b8a94753&vid=56ff02c19ce1371a2a49ed5e7fc9af49b8a94753&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR