2-Methylcinnamic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M1295-25G
M1295-5G
TCM1295-025GEA
309.05
CAD
TCM1295-025G
TCM1295-005G
2-Methylcinnamic acid ≥98.0% (by GC, titration analysis)
2-Methylcinnamic acid
CAS Number: 2373-76-4
MDL Number: MFCD00016841
Molecular Formula: C10H10O2
Molecular Weight: 162.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 176
Formula:
C₁₀H₁₀O₂ Boiling Pt: 303 °C (1013 hPa) Melting Pt: 176 °C |
MDL Number:
MFCD00016841 CAS Number: 2373-76-4 EINECS: 219-150-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...