5-Methoxy-2-methylbenzothiazole ≥98.0%

Supplier: TCI America

M1281-5G M1281-25G
TCM1281-005GEA 137.46 CAD
TCM1281-005G TCM1281-25G
5-Methoxy-2-methylbenzothiazole ≥98.0%
5-Methoxy-2-methylbenzothiazole

CAS Number: 2941-69-7
MDL Number: MFCD00082664
Molecular Formula: C9H9NOS
Molecular Weight: 179.24
Purity/Analysis Method: >98.0% (GC)
Form: Lump
Boiling point (°C): 135
Melting point (°C): 37

Formula: C₉H₉NOS
MW: 179.24 g/mol
Boiling Pt: 135 °C
Melting Pt: 37 °C
MDL Number: MFCD00082664
CAS Number: 2941-69-7
EINECS: 220-929-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d567a36c03e2a211e852cf38b5cb1e9dbea0a40&vid=9d567a36c03e2a211e852cf38b5cb1e9dbea0a40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR