Sodium methanesulfinate ≥90.0% (by titrimetric analysis)
Supplier: TCI America
M1255-25G
M1255-5G
TCM1255-025GEA
380.78
CAD
TCM1255-025G
TCM1255-005G
Sodium methanesulfinate ≥90.0% (by titrimetric analysis)
Sodium methanesulphinate
CAS Number: 20277-69-4
MDL Number: MFCD00040392
Molecular Formula: CH4O2S
Molecular Weight: 102.08
Purity/Analysis Method: >90.0% (T)
Form: Crystal
Melting point (°C): 226
Other Product Names / Synonyms: Methanesulfinic Acid Sodium Salt
Formula:
CH₃NaO₂S Melting Pt: 226 °C |
MDL Number:
MFCD00040392 CAS Number: 20277-69-4 EINECS: 243-669-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...