4-Methoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Methoxybenzeneboronic acid, Dihydroxy(4-methoxyphenyl)borane, (4-Methoxyphenyl)metaboric acid

M1252-5G M1252-25G
TCM1252-005GEA 114.8 CAD
TCM1252-005G TCM1252-025G
4-Methoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Methoxyphenylboronic acid

CAS Number: 5720-07-0
MDL Number: MFCD00039139
Molecular Formula: C7H9BO3
Molecular Weight: 151.96
Form: Crystal
Color: White


Other Product Names / Synonyms: 4-Methoxybenzeneboronic Acid

Formula: C₇H₉BO₃
MW: 151.96 g/mol
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00039139
CAS Number: 5720-07-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=71d7d36b7608726cd802fea10658e3e682001667&vid=71d7d36b7608726cd802fea10658e3e682001667&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR