4-Methoxyindole ≥98.0%

Supplier: TCI America
Warning

Synonyms: 4-Methoxy-1H-indole, Indol-4-yl methyl ether

M1250-5G M1250-1G
TCM1250-005GEA 124.82 CAD
TCM1250-005G TCM1250-001G
4-Methoxyindole ≥98.0%
4-Methoxyindole

CAS Number: 4837-90-5
MDL Number: MFCD00009737
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 181
Melting point (°C): 70

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 181 °C
Melting Pt: 70 °C
MDL Number: MFCD00009737
CAS Number: 4837-90-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f22db203231fc23d2d61e8fe76a2ec89b4bad43&vid=8f22db203231fc23d2d61e8fe76a2ec89b4bad43&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR