4-Methylbenzophenone ≥95.0%

Supplier: TCI America

Synonyms: p-Methylbenzophenone, Phenyl(p-tolyl)methanone

M1248-25G M1248-250G
TCM1248-025GEA 44.98 CAD
TCM1248-025G TCM1248-250G
4-Methylbenzophenone ≥95.0%
4-Methylbenzophenone

CAS Number: 134-84-9
MDL Number: MFCD00008553
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 328
Melting point (°C): 56
Flash Point (°C): 143

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
Boiling Pt: 328 °C
Melting Pt: 56 °C
Density: 0.9926 g/cm³ (20 °C)
Flash Pt: 143 °C
MDL Number: MFCD00008553
CAS Number: 134-84-9
EINECS: 205-159-1
Merck Index: 12,07470
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb005239a4a60d9e8505ba0b44f1e4e9dd119a06&vid=cb005239a4a60d9e8505ba0b44f1e4e9dd119a06&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR