3-Methylbenzophenone ≥95.0%

Supplier: TCI America

Synonyms: m-Methylbenzophenone

M1247-5G M1247-25G
TCM1247-005GEA 49.82 CAD
TCM1247-005G TCM1247-025G
3-Methylbenzophenone ≥95.0%
3-Methylbenzophenone

CAS Number: 643-65-2
MDL Number: MFCD00008535
Molecular Formula: C14H12O
Molecular Weight: 196.25
Purity/Analysis Method: >95.0% (T)
Form: Clear Liquid
Boiling point (°C): 316
Flash Point (°C): 113
Specific Gravity (20/20): 1.09

Formula: C₁₄H₁₂O
Boiling Pt: 316 °C
Melting Pt: 0…2 °C
Density: 1.09 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008535
CAS Number: 643-65-2
EINECS: 211-401-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b627b855f8d37162f865e7cfd670a10ddfc1c9be&vid=b627b855f8d37162f865e7cfd670a10ddfc1c9be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR