2-Methylbiphenyl ≥95.0%

Supplier: TCI America
Warning

Synonyms: 2-Phenyltoluene

M1242-5G M1242-1G
TCM1242-005GEA 295.46 CAD
TCM1242-005G TCM1242-1G
2-Methylbiphenyl ≥95.0%
2-Methylbiphenyl

CAS Number: 643-58-3
MDL Number: MFCD00008517
Molecular Formula: C13H12
Molecular Weight: 168.24
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 256
Flash Point (°C): 107
Specific Gravity (20/20): 1.01


Other Product Names / Synonyms: 2-Phenyltoluene

Formula: C₁₃H₁₂
MW: 168.24 g/mol
Boiling Pt: 256 °C
Melting Pt: –0.2 °C
Density: 1.01 (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00008517
CAS Number: 643-58-3
EINECS: 211-400-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dc7a53a88557503cdd1ec1f6c747bac8bca89e37&vid=3837bb105a5d69b45b086deb3fcdc1af7817e33b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR