6-Methoxyindole ≥98.0%

Supplier: TCI America
Warning

M1214-100MG M1214-1G
TCM1214-100MGEA 42.66 CAD
TCM1214-100MG TCM1214-1G
6-Methoxyindole ≥98.0%
6-Methoxyindole

CAS Number: 3189-13-7
MDL Number: MFCD00022780
Molecular Formula: C9H9NO
Molecular Weight: 147.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 93

Formula: C₉H₉NO
Melting Pt: 93 °C
Storage Temperature: Freezer
MDL Number: MFCD00022780
CAS Number: 3189-13-7
EINECS: 221-689-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=434bbdb853ddec1d2288f61d211531fb4daf8502&vid=434bbdb853ddec1d2288f61d211531fb4daf8502&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR