2-Fluoromesitylene ≥98.0%

Supplier: TCI America

Synonyms: 2-Fluoro-1,3,5-trimethylbenzene, Fluoromesitylene, 2,4,6-Trimethylfluorobenzene

M1201-5G M1201-1G
TCM1201-5GEA 284.4 CAD
TCM1201-5G TCM1201-001G
2-Fluoromesitylene ≥98.0%
2-Fluoromesitylene

CAS Number: 392-69-8
MDL Number: MFCD00052365
Molecular Formula: C9H11F
Molecular Weight: 138.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Specific Gravity (20/20): 0.98


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoromesitylene, 2-Fluoro-1,3,5-trimethylbenzene

Formula: C₉H₁₁F
MW: 138.18 g/mol
Boiling Pt: 165 °C
Density: 0.98 (20 °C)
MDL Number: MFCD00052365
CAS Number: 392-69-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9eb1f31ccc58e4047a949e7af5217c2df4ab2baa&vid=9eb1f31ccc58e4047a949e7af5217c2df4ab2baa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR