4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98.0% (by GC, titration analysis) free radical

Supplier: TCI America

M1197-1G M1197-5G
TCM1197-1GEA 85.32 CAD
TCM1197-1G TCM1197-005G
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98.0% (by GC, titration analysis) free radical
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-Methoxy-TEMPO, free radical)

[Catalyst for Oxidation]
CAS Number: 95407-69-5
MDL Number: MFCD00270334
Molecular Formula: C10H20NO2
Molecular Weight: 186.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 43


Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Methoxy-TEMPO

Formula: C₁₀H₂₀NO₂
Melting Pt: 43 °C
MDL Number: MFCD00270334
CAS Number: 95407-69-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e874dbce343de19d8443edc4ce4fcd30c854c40&vid=015b9fd6bd585252e8a2052ea1ccf5e067202366&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR