3-Methyl-2-nitroanisole ≥98.0%

Supplier: TCI America
Warning

M1196-5G M1196-25G
TCM1196-5GEA 80.41 CAD
TCM1196-5G TCM1196-025G
3-Methyl-2-nitroanisole ≥98.0%
3-Methyl-2-nitroanisole

CAS Number: 5345-42-6
MDL Number: MFCD00007179
Molecular Formula: C8H9NO3
Molecular Weight: 167.16
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 170
Melting point (°C): 49
Flash Point (°C): 113


Other Product Names / Synonyms: 1-Methoxy-3-methyl-2-nitrobenzene, 3-Methyl-2-nitroanisole

Formula: C₈H₉NO₃
Boiling Pt: 170 °C
Melting Pt: 49 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007179
CAS Number: 5345-42-6
EINECS: 226-295-8
UN: 2446
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d7d5ce7ad574e421cd50fb98d8a73658f8b1a088&vid=0d2a98a2c118f94d11fad0fb7fb1275b4640176f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR