N-Phenyl-p-toluidine ≥98.0%
Supplier: TCI America
M1173-1G
M1173-5G
TCM1173-001GEA
33.18
CAD
TCM1173-001G
TCM1173-005G
N-Phenyl-p-toluidine ≥98.0%
N-Phenyl-p-toluidine
CAS Number: 620-84-8
MDL Number: MFCD00092921
Molecular Formula: C13H13N
Molecular Weight: 183.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 317
Melting point (°C): 90
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Diphenylamines (for High-Performance Polymer Research)
Other Product Names / Synonyms: N-Phenyl-p-toluidine
Formula:
C₁₃H₁₃N Boiling Pt: 317 °C Melting Pt: 90 °C |
MDL Number:
MFCD00092921 CAS Number: 620-84-8 EINECS: 210-655-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...