4-Methoxybenzyl mercaptan ≥98.0%

Supplier: TCI America

M1165-5G M1165-25G
TCM1165-005GEA 82.16 CAD
TCM1165-005G TCM1165-025G
4-Methoxybenzyl mercaptan ≥98.0%
4-Methoxybenzyl mercaptan

CAS Number: 6258-60-2
MDL Number: MFCD00004871
Molecular Formula: C8H10OS
Molecular Weight: 154.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 126
Flash Point (°C): 110
Specific Gravity (20/20): 1.12


Other Product Names / Synonyms: 4-Methoxybenzyl Mercaptan

Formula: C₈H₁₀OS
Boiling Pt: 126 °C
Density: 1.12 (20 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004871
CAS Number: 6258-60-2
EINECS: 228-393-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4bc62e864cfb13a31f4eaa75743590f17099479f&vid=4bc62e864cfb13a31f4eaa75743590f17099479f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR