Methyl phenyl sulfoxide ≥98.0%

Supplier: TCI America

M1148-25G M1148-5G
TCM1148-25GEA 165.9 CAD
TCM1148-25G TCM1148-005G
Methyl phenyl sulfoxide ≥98.0%
Methyl phenyl sulphoxide

CAS Number: 1193-82-4
MDL Number: MFCD00002088
Molecular Formula: C7H8OS
Molecular Weight: 140.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 86
Freezing point (°C): 29

Formula: C₇H₈OS
Flash Pt: 86 °C
MDL Number: MFCD00002088
CAS Number: 1193-82-4
EINECS: 214-781-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b482c615097ecd466bc84ecada80db330d08373&vid=5b482c615097ecd466bc84ecada80db330d08373&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR