2-Methyl-1,3-cyclohexanedione ≥98.0%

Supplier: TCI America

Synonyms: 2-Methylcyclohexane-1,3-dione

M1133-25G M1133-5G
TCM1133-025GEA 450.3 CAD
TCM1133-025G TCM1133-005G
2-Methyl-1,3-cyclohexanedione ≥98.0%
2-Methyl-1,3-cyclohexanedione

CAS Number: 1193-55-1
MDL Number: MFCD00001587
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal

Formula: C₇H₁₀O₂
Boiling Pt: 100…102 °C (8 mmHg)
Melting Pt: 206…209 °C
Density: 1.313 g/cm³ (–150.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001587
CAS Number: 1193-55-1
EINECS: 214-773-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5bbd4d6f9e430380d0322a97f4824d5508a1bf40&vid=5bbd4d6f9e430380d0322a97f4824d5508a1bf40&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR