6-Methoxy-2-naphthaldehyde ≥97.0%
Supplier: TCI America
M1090-25G
M1090-5G
TCM1090-025GEA
180.12
CAD
TCM1090-025G
TCM1090-5G
6-Methoxy-2-naphthaldehyde ≥97.0%
6-Methoxy-2-naphthaldehyde
CAS Number: 3453-33-6
MDL Number: MFCD00081152
Molecular Formula: C12H10O2
Molecular Weight: 186.21
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 196
Melting point (°C): 82
Formula:
C₁₂H₁₀O₂ Boiling Pt: 196 °C Melting Pt: 82 °C |
MDL Number:
MFCD00081152 CAS Number: 3453-33-6 EINECS: 222-377-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...