Dibenzyl malonate ≥95.0%

Supplier: TCI America

Synonyms: Malonic acid dibenzyl ester, 1,3-Propanedioic acid dibenzyl ester

M1080-25G M1080-500G M1080-100G
TCM1080-025GEA 164.32 CAD
TCM1080-025G TCM1080-500G TCM1080-100G
Dibenzyl malonate ≥95.0%
Dibenzyl malonate

CAS Number: 15014-25-2
MDL Number: MFCD00004779
Molecular Formula: C17H16O4
Molecular Weight: 284.31
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 234
Flash Point (°C): 110
Specific Gravity (20/20): 1.17


Other Product Names / Synonyms: Malonic Acid Dibenzyl Ester

Formula: C₁₇H₁₆O₄
MW: 284.31 g/mol
Boiling Pt: 188
Density: 1.17 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00004779
CAS Number: 15014-25-2
EINECS: 239-099-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=196f53065787ee5d3d569aaeeacfdd22f1dc2a26&vid=196f53065787ee5d3d569aaeeacfdd22f1dc2a26&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR