3-Methyl-2-cyclohexen-1-one ≥98.0%

Supplier: TCI America

M1043-25G M1043-5G
TCM1043-025GEA 145.36 CAD
TCM1043-025G TCM1043-5G
3-Methyl-2-cyclohexen-1-one ≥98.0%
3-Methyl-2-cyclohexen-1-one

CAS Number: 1193-18-6
MDL Number: MFCD00001581
Molecular Formula: C7H10O
Molecular Weight: 110.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Flash Point (°C): 68
Specific Gravity (20/20): 0.97

Formula: C₇H₁₀O
Boiling Pt: 197 °C
Melting Pt: 198…199 °C
Density: 0.97 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00001581
CAS Number: 1193-18-6
EINECS: 214-769-7
UN: 1224
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f7f775eecfe967afccd0d2be4c7e8348538d7f46&vid=f7f775eecfe967afccd0d2be4c7e8348538d7f46&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR