2-Nitro-m-anisic acid ≥99.0%
Supplier: TCI America
Synonyms:
3-Methoxy-2-nitrobenzoic acid
M1036-10G
M1036-1G
TCM1036-010GEA
391.84
CAD
TCM1036-010G
TCM1036-001G
2-Nitro-m-anisic acid ≥99.0%
2-Nitro-m-anisic acid
CAS Number: 4920-80-3
MDL Number: MFCD00007143
Molecular Formula: C8H7NO5
Molecular Weight: 197.15
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Other Product Names / Synonyms: 2-Nitro-m-anisic Acid
Formula:
C₈H₇NO₅ Boiling Pt: 388 °C (1013 hPa) Melting Pt: 174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007143 CAS Number: 4920-80-3 EINECS: 225-549-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...