6-Nitro-o-toluic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1002-25G M1002-10G
TCM1002-25GEA 295.46 CAD
TCM1002-25G TCM1002-010G
6-Nitro-o-toluic acid ≥98.0% (by titrimetric analysis)
6-Nitro-o-toluic acid

CAS Number: 13506-76-8
MDL Number: MFCD00007267
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 156


Other Product Names / Synonyms: 6-Nitro-o-toluic Acid

Formula: C₈H₇NO₄
Boiling Pt: 353 °C (1013 hPa)
Melting Pt: 156 °C
MDL Number: MFCD00007267
CAS Number: 13506-76-8
EINECS: 236-833-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57c251c816d3900ca82f302cd32a95634579a9d1&vid=3d442566f83e9620ffcedd0c80e9a9af2238601e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR