3-Nitro-p-toluic acid ≥99.0%
Supplier: TCI America
Synonyms:
4-Methyl-3-nitrobenzoic acid
M0950-500G
M0950-25G
TCM0950-500GEA
192.76
CAD
TCM0950-500G
TCM0950-025G
3-Nitro-p-toluic acid ≥99.0%
3-Nitro-p-toluic acid
CAS Number: 96-98-0
MDL Number: MFCD00007174
Molecular Formula: C8H7NO4
Molecular Weight: 181.15
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 190
Other Product Names / Synonyms: 3-Nitro-p-toluic Acid
Formula:
C₈H₇NO₄ Boiling Pt: 360 °C (1013 hPa) Melting Pt: 190 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007174 CAS Number: 96-98-0 EINECS: 202-549-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...