5-Methyl-1,3-cyclohexanedione ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
M0945-5G
M0945-1G
TCM0945-005GEA
85.32
CAD
TCM0945-005G
TCM0945-001G
5-Methyl-1,3-cyclohexanedione ≥98.0% (by GC, titration analysis)
5-Methyl-1,3-cyclohexanedione
CAS Number: 4341-24-6
MDL Number: MFCD00010379
Molecular Formula: C7H10O2
Molecular Weight: 126.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 127
Formula:
C₇H₁₀O₂ Melting Pt: 127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010379 CAS Number: 4341-24-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...