4-Methoxybiphenyl ≥99.0%

Supplier: TCI America

M0944-5G M0944-25G
TCM0944-005GEA 44.24 CAD
TCM0944-005G TCM0944-025G
4-Methoxybiphenyl ≥99.0%
4-Methoxybiphenyl

CAS Number: 613-37-6
MDL Number: MFCD00014897
Molecular Formula: C13H12O
Molecular Weight: 184.24
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 157
Melting point (°C): 89


Other Product Names / Synonyms: 4-Phenylanisole

Formula: C₁₃H₁₂O
Boiling Pt: 157 °C
Melting Pt: 89 °C
MDL Number: MFCD00014897
CAS Number: 613-37-6
EINECS: 210-339-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87e47e61a84e8c664e58a2ee3daaf0ffec470686&vid=87e47e61a84e8c664e58a2ee3daaf0ffec470686&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR