4-Hydroxy-1-methylpiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M0933-500G M0933-25G
TCM0933-500GEA 350.76 CAD
TCM0933-500G TCM0933-25G
4-Hydroxy-1-methylpiperidine ≥98.0% (by GC, titration analysis)
4-Hydroxy-1-methylpiperidine

CAS Number: 106-52-5
MDL Number: MFCD00006500
Molecular Formula: C6H13NO
Molecular Weight: 115.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 105
Flash Point (°C): 87
Freezing point (°C): 27


Other Product Names / Synonyms: 1-Methyl-4-piperidinol

Formula: C₆H₁₃NO
Boiling Pt: 105 °C
Melting Pt: 29…31 °C
Density: 0.98 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00006500
CAS Number: 106-52-5
EINECS: 203-406-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ccaeefb8c9ec5f15c4a756debc13609aee5bf45&vid=2ccaeefb8c9ec5f15c4a756debc13609aee5bf45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR