3-Methyl-2-phenylpyridine ≥98.0%
Supplier: TCI America
M0932-5G
M0932-25G
TCM0932-5GEA
188.02
CAD
TCM0932-5G
TCM0932-25G
3-Methyl-2-phenylpyridine ≥98.0%
3-Methyl-2-phenylpyridine
CAS Number: 10273-90-2
MDL Number: MFCD00006283
Molecular Formula: C12H11N
Molecular Weight: 169.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 96
Specific Gravity (20/20): 1.07
Product Categories: Materials Chemistry, Ligands for Functional Metal Complexes
Other Product Names / Synonyms: 2-Phenyl-beta-picoline
Formula:
C₁₂H₁₁N Boiling Pt: 160 °C Density: 1.07 (20 °C) Flash Pt: 96 °C |
MDL Number:
MFCD00006283 CAS Number: 10273-90-2 EINECS: 233-619-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...