L(-)-Methioninol ≥98.0%
Supplier: TCI America
Synonyms:
(S)-2-Amino-4-(methylthio)butan-1-ol, L-Methioninol
CAS Number: 2899-37-8
MDL Number: MFCD00004735
Molecular Formula: C5H13NOS
Molecular Weight: 135.23
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 35
Specific rotation [a]20/D: -14 deg (C=1.4, EtOH)
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-2-Amino-4-methylthio-1-butanol
Formula:
C₅H₁₃NOS MW: 135.2 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD00004735 CAS Number: 2899-37-8 EINECS: 220-788-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...