2-Methyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methylcyclopentane-1,3-dione

M0901-5G M0901-25G
TCM0901-005GEA 47.4 CAD
TCM0901-005G TCM0901-25G
2-Methyl-1,3-cyclopentanedione ≥97.0% (by GC, titration analysis)
2-Methyl-1,3-cyclopentanedione

CAS Number: 765-69-5
MDL Number: MFCD00001406
Molecular Formula: C6H8O2
Molecular Weight: 112.13
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 214

Formula: C₆H₈O₂
MW: 112.13 g/mol
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: 214 °C
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001406
CAS Number: 765-69-5
EINECS: 212-153-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11bb2f3e0dddeac7bbdff10851747aef8c559530&vid=11bb2f3e0dddeac7bbdff10851747aef8c559530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR