3-Methylthiophene-2-carboxaldehyde ≥85.0%
Supplier: TCI America
M0899-25G
M0899-100G
TCM0899-025GEA
72.68
CAD
TCM0899-025G
TCM0899-100G
3-Methylthiophene-2-carboxaldehyde ≥85.0%
3-Methylthiophene-2-carboxaldehyde
CAS Number: 5834-16-2
MDL Number: MFCD00005430
Molecular Formula: C6H6OS
Molecular Weight: 126.17
Purity/Analysis Method: >85.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 82
Specific Gravity (20/20): 1.19
Other Product Names / Synonyms: 2-Formyl-3-methylthiophene
Formula:
C₆H₆OS Boiling Pt: 113 °C Density: 1.19 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00005430 CAS Number: 5834-16-2 EINECS: 227-418-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...