4-Methoxy-1-naphthol ≥98.0%

Supplier: TCI America

Synonyms: 4-Methoxynaphthalen-1-ol, 1-Hydroxy-4-methoxynaphthalene

M0854-5G M0854-25G
TCM0854-005GEA 138.88 CAD
TCM0854-005G TCM0854-025G
4-Methoxy-1-naphthol ≥98.0%
4-Methoxy-1-naphthol

CAS Number: 84-85-5
MDL Number: MFCD00003976
Molecular Formula: C11H10O2
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 130


Other Product Names / Synonyms: 1-Hydroxy-4-methoxynaphthalene

Formula: C₁₁H₁₀O₂
MW: 174.2 g/mol
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 130 °C
MDL Number: MFCD00003976
CAS Number: 84-85-5
EINECS: 201-566-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f703859040453728f65f94816f667a2481d4ebe&vid=0f703859040453728f65f94816f667a2481d4ebe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR