(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(2R)-(+)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid, (R)-3,3,3-Trifluoro-2-methoxy-2-phenylpropanoic acid
[Optical Resolving]
CAS Number: 20445-31-2
MDL Number: MFCD00004184
Molecular Formula: C10H9F3O3
Molecular Weight: 234.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Flash Point (°C): 112
Freezing point (°C): 35
Specific rotation [a]20/D: 72 deg (C=2, MeOH)
Product Categories: Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (+)-Mosher's Acid, (+)-MTPA
Formula:
C₁₀H₉F₃O₃ MW: 234.17 g/mol Flash Pt: 112 °C |
MDL Number:
MFCD00004184 CAS Number: 20445-31-2 EINECS: 243-829-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...