(R)-(-)-α-Methoxyphenylacetic acid ≥98.0%

Supplier: TCI America
Warning

Synonyms: (R)-(-)-alpha-Methoxyphenylacetic Acid

M0830-5G M0830-1G M0830-100MG
TCM0830-5GEA 197.69 CAD
TCM0830-5G TCM0830-1G TCM0830-100MG
(R)-(-)-α-Methoxyphenylacetic acid ≥98.0%
(R)-(-)-α-Methoxyphenylacetic acid

CAS Number: 3966-32-3
MDL Number: MFCD00004250
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 68
Specific rotation [a]20/D: -147 deg (C=0.5, EtOH)

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 130 °C
Melting Pt: 68 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004250
CAS Number: 3966-32-3
EINECS: 223-580-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1e61408107d45026a9fa2e9fc889ee01365237e5&vid=1e61408107d45026a9fa2e9fc889ee01365237e5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR