(S)-(+)-α-Methoxyphenylacetic acid ≥98.0%
Supplier: TCI America
|
Warning
|
Synonyms:
(S)-(+)-α-Methoxyphenylacetic acid, (S)-alpha-Methoxyphenylacetic Acid
M0829-5G
M0829-1G
TCM0829-5GEA
496.78
CAD
TCM0829-5G
TCM0829-001G
(S)-(+)-α-Methoxyphenylacetic acid ≥98.0%
(S)-(+)-α-Methoxyphenylacetic acid
CAS Number: 26164-26-1
MDL Number: MFCD00064216
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 66
Specific rotation [a]20/D: 147 deg (C=0.5, EtOH)
Formula:
C₉H₁₀O₃ MW: 166.18 g/mol Boiling Pt: 130 °C Melting Pt: 66 °C |
MDL Number:
MFCD00064216 CAS Number: 26164-26-1 EINECS: 247-492-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...