(Methoxymethyl)triphenylphosphonium chloride ≥99.0% (by HPLC)

Supplier: TCI America
Warning

M0828-25G M0828-500G M0828-100G
TCM0828-025GEA 52.44 CAD
TCM0828-025G TCM0828-500G TCM0828-100G
(Methoxymethyl)triphenylphosphonium chloride ≥99.0% (by HPLC)
(Methoxymethyl)triphenylphosphonium chloride

CAS Number: 4009-98-7
MDL Number: MFCD00011800
Molecular Formula: C20H20ClOP
Molecular Weight: 342.80
Purity/Analysis Method: >99.0% (HPLC)
Form: Crystal
Melting point (°C): 202

Formula: C₂₀H₂₀ClOP
Melting Pt: 202 °C
MDL Number: MFCD00011800
CAS Number: 4009-98-7
EINECS: 223-664-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db87a82e564b8cd1add974988aae30723140ffb8&vid=db87a82e564b8cd1add974988aae30723140ffb8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR