2-(Methylamino)pyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M0823-500G
M0823-25G
TCM0823-500GEA
720.48
CAD
TCM0823-500G
TCM0823-025G
2-(Methylamino)pyridine ≥98.0% (by GC, titration analysis)
2-(Methylamino)pyridine
CAS Number: 4597-87-9
MDL Number: MFCD00006250
Molecular Formula: C6H8N2
Molecular Weight: 108.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 201
Flash Point (°C): 102
Freezing point (°C): 18
Specific Gravity (20/20): 1.06
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 201 °C Melting Pt: 15 °C Density: 1.06 (20 °C) Flash Pt: 102 °C |
MDL Number:
MFCD00006250 CAS Number: 4597-87-9 EINECS: 224-997-9 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...