1H-1,2,4-Triazole-3-thiol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0814-25G M0814-500G M0814-100G
TCM0814-025GEA 71.1 CAD
TCM0814-025G TCM0814-500G TCM0814-100G
1H-1,2,4-Triazole-3-thiol ≥98.0% (by GC, titration analysis)
1H-1,2,4-Triazole-3-thiol

CAS Number: 3179-31-5
MDL Number: MFCD00005229
Molecular Formula: C2H3N3S
Molecular Weight: 101.13
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 218


Other Product Names / Synonyms: 1,2,4-Triazole-3-thiol

Formula: C₂H₃N₃S
Melting Pt: 218 °C
MDL Number: MFCD00005229
CAS Number: 3179-31-5
EINECS: 221-656-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cefe2c3664fa86008cfb4123040760bbe65f072&vid=0cefe2c3664fa86008cfb4123040760bbe65f072&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR