4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M0790-25G M0790-250G
TCM0790-025GEA 116.92 CAD
TCM0790-025G TCM0790-250G
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) ≥98.0% (by HPLC, titration analysis)
4-Methoxytrityl chloride (p-Anisylchlorodiphenylmethane)

[Hydroxyl Protecting Agent]
CAS Number: 14470-28-1
MDL Number: MFCD00000814
Molecular Formula: C20H17ClO
Molecular Weight: 308.81
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 124
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis), Nucleic Acid Synthesis
Other Product Names / Synonyms: 4-Monomethoxytrityl Chloride

Formula: C₂₀H₁₇ClO
Melting Pt: 124 °C
MDL Number: MFCD00000814
CAS Number: 14470-28-1
EINECS: 238-463-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2e7d4e433ce88b18d8875d47a702216ae0790d0&vid=46f043f05227befa6b362830619cc148c1d1eb84&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR