6-Methoxy-1-tetralone ≥99.0%

Supplier: TCI America

M0787-250G M0787-25G
TCM0787-250GEA 534.07 CAD
TCM0787-250G TCM0787-025G
6-Methoxy-1-tetralone ≥99.0%
6-Methoxy-1-tetralone

CAS Number: 1078-19-9
MDL Number: MFCD00001695
Molecular Formula: C11H12O2
Molecular Weight: 176.22
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: 3,4-Dihydro-6-methoxy-1(2H)-naphthalenone

Formula: C₁₁H₁₂O₂
Melting Pt: 79 °C
MDL Number: MFCD00001695
CAS Number: 1078-19-9
EINECS: 214-078-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07d467590cc072f5ed015969d110d68e71b32425&vid=07d467590cc072f5ed015969d110d68e71b32425&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR